missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Mouse Lysyl Oxidase/LOX (NP_034858) VersaClone cDNA

Artikelnummer. 18759837
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18759837 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18759837 Leverandør R&D Systems™ Leverandørnr. RDC2688

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • Also known as: rrg; TSC-160; AI893619
  • LOX belongs to the lysyl oxidase family of proteins.
  • The secreted proprotein is proteolytically processed to an active mature peptide and a propeptide.
  • This propeptide is thought to function in tumor suppression by inhibiting the Ras signaling pathway.
  • The active enzyme plays a role in cross-linking of collagen and elastin and is essential for development of cardiovascular and respiratory systems, and development of skin and connective tissue.
  • Alternatively spliced transcripts encoding different proteins have been described.
  • The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Tekniske data

Gene mLox
Adgangsnummer NP_034858
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl, pH 8.5
Indsæt type 1249 bp
Gen-id (Entrez) 4015
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Mouse
Mængde 10 μg
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.