missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Mouse Leptin R (NP_666258) VersaClone cDNA

Artikelnummer. 18726798
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18726798 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18726798 Leverandør R&D Systems™ Leverandørnr. RDC2183

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

R&D Systems™ Mouse Leptin R (NP_666258) VersaClone cDNA belongs to the gp130 family of cytokine receptors that are known to stimulate gene transcription via activation of cytosolic STAT proteins.

The Mouse Leptin R (NP_666258) VersaClone cDNA clone contains the complete ORF for Leptin R, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Leptin R insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Tekniske data

Gene mLepr
Adgangsnummer NP_666258
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl
Indsæt type 3502bp
pH 8.5
Gen-id (Entrez) 3953
Indhold og opbevaring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Arter Mouse
Mængde 10 μg
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.