missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Mouse IL-17/IL-17A Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 17 publications
Brand: R&D Systems MAB721-500
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Tekniske data
| IL-17/IL-17A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3605 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, ELISA | |
| 50101 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q62386 | |
| IL17A | |
| E. coli-derived recombinant mouse IL-17 Thr22-Ala158 Accession # Q62386 | |
| 500 μg | |
| Primary | |
| Detects mouse IL-17 in ELISAs and Western blots. In direct ELISAs, 100% reactivity with recombinant mouse (rm) IL-17A/IL-17F heterodimer, 50% cross-reactivity with recombinant human IL-17, and 25% cross-reactivity with recombinant canine IL-17 is observed. No cross-reactivity with rmIL-17B, rmIL-17C, rmIL-17D, rmIL-17E, or rmIL-17F is observed. In Western blots, cross-reactivity with recombinant rat IL-17A is observed. | |
| Mouse | |
| Supernatant |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion