missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ Mouse IgG2a kappa Isotype Control (eBM2a), Super Bright™ 600, eBioscience™

Artikelnummer. 16301494
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100μ g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16301494 100μ g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16301494 Leverandør Invitrogen™ Leverandørnr. 63472482

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Isotype Control

This is a monoclonal mouse IgG2a antibody. It is used as an isotype control for mouse IgG2a antibodies. Applications Reported: This eBM2a antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBM2a antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse spleen cells. Use isotype control at the same concentration as experimental antibody. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 2-8°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorochrome performance after fixation can be made, but clone specific pe...

Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY

Especificaciones

Antigen Mouse IgG2a kappa
Applikationer Control, Flow Cytometry
Klassifikation Monoclonal
Klon eBM2a
Koncentration 0.2 mg/mL
Konjugeret Super Bright 600
Formulering PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Værtsarter Mus
Oprensningsmetode Affinity chromatography
Mængde 100μ g
Regulatorisk status RUO
Primær eller sekundær Isotype Control (Primary)
Målarter Not Applicable
Indhold og opbevaring 4°C, store in dark, DO NOT FREEZE!
Produkttype Isotype Control
Form Væske
Isotype IgG2a κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.