Få mere at vide
Mouse IgG2A Fluorescein-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems IC003F
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG2a Monoclonal specifically detects IgG2a in Mouse samples. It is validated for Control.
Tekniske data
| IgG2a | |
| 20102 | |
| Control 10 uL/10^6 cells | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Selected for use as mouse IgG 2A isotype control for flow cytometry applications. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| KLH | |
| 200 Tests | |
| Mouse | |
| IgG2a |