Få mere at vide
Mouse IgG1 PerCP-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC002C
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
IgG1 Monoclonal specifically detects IgG1 in Mouse samples. It is validated for Control.
Tekniske data
| IgG1 | |
| 11711 | |
| Control 10 uL/10^6 cells | |
| IgG1, Igh-4, RGD1359539, VH7183 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Selected for use as mouse IgG 1 isotype control for flow cytometry applications. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Monoclonal | |
| PerCP | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IGHG1 | |
| KLH | |
| 200 Tests | |
| Mouse | |
| IgG1 κ |