missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgG1 Isotype Control, PerCP, Invitrogen™
Mouse Isotype Control
Brand: Invitrogen™ MG131
Additional Details : Weight : 1.00000kg
Description
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Flow Cytometry.Specifications
Mouse IgG1 | |
Monoclonal | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Isotype Control (Primary) | |
2-8°C | |
IgG1 |
Flow Cytometry | |
PerCP | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
purified | |
ASR | |
Not Applicable | |
Liquid |