Learn More
Invitrogen™ Mouse IgG1 Isotype Control, Alexa Fluor™ 488, Invitrogen™
Mouse Isotype Control
Brand:  Invitrogen™ MG120
Additional Details : Weight : 0.50000kg
Product Code. 10058072
  • 1232.00 DKK / 500µL
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
Mouse IgG1 Isotype Control for Flow
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
Specifications
Specifications
Mouse IgG1
Alexa Fluor 488
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
purified
ASR
Isotype Control (Primary)
2-8°C
IgG1
Wet Ice
Flow Cytometry
PBS with 0.1% sodium azide
Mouse
500 μL
Whole Antibody
Not Applicable
Liquid
50 Tests
Alexa Fluor™