Få mere at vide
R&D Systems™ Mouse HB-EGF DuoSet ELISA
Artikelnummer. 30021248
Format:
96-well Plates
Artikelnummer. 30021248
Brand: R&D Systems™ DY823905
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Mouse HB-EGF DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Mouse HB-EGF DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse HB-EGF.
TRUSTED_SUSTAINABILITY
Tekniske data
Produkttype ELISA Kit
Arter Mouse
Bindende mål HB-EGF
Format 96-well Plates
Til brug med (applikation) ELISA
Gen-id (Entrez) 1839
Prøvetype Cell Culture Supernatant, Serum, Plasma
Indhold og opbevaring Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Research Use Only.