missing translation for 'onlineSavingsMsg'
Få mere at vide

Gibco™ Mouse GM-CSF Recombinant Protein, PeproTech®

Artikelnummer. 17842913
Klik for at se tilgængelige muligheder
Mængde:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Pakningsstørrelse:
100µg
20µg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 17842913

Brand: Gibco™ 3150350UG

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Edge
Denne vare kan ikke returneres. Se returpolicy

Recombinant Protein

Recombinant Murine GM-CSF is a 14.2 kDa globular protein consisting of 125 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The GM-CSF gene encodes for a cytokine that regulates the production, differentiation, and function of macrophages and granulocytes. The active form of the protein exists as a homodimer in the extracellular space. This gene is located in a cluster of related genes at the chromosome region 5q31, which has been associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include interleukins 4, 5, and 13. The gene is involved in promoting tissue inflammation. Elevated levels of cytokines, including the one produced by this gene, have been observed in SARS-CoV-2 infected patients with acute respiratory distress syndrome. Mice that lack this gene or its receptor have been shown to develop pulmonary alveolar proteinosis.
TRUSTED_SUSTAINABILITY

Tekniske data

Adgangsnummer P01587
Til brug med (applikation) ELISA, Functional Assay, Western Blot
Formulering protein with no preservative
Gen-id (Entrez) 12981
Molekylvægt (g/mol) 14.2 kDa
Navn Mouse GM-CSF
Mængde 50 μg
Kilde E. coli
Regulatorisk status RUO
Endotoksin koncentration <1 EU/ μg
Gene Alias colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; molgramostin; put. GM-CSF; sargramostim
Fælles navn GM-CSF
Gen symbol CSF2
Biologisk aktivitet The ED50 as determined by the dose-dependent stimulation of the proliferation of murine FDC-P1 cells is ≤ 0.05 ng/ml, corresponding to a specific activity of ≥ 2 x 10^7 units/mg
Konjugeret Unconjugated
Rekombinant Recombinant
Sekvens MAPTRSPITV TRPWKHVEAI KEALNLLDDM PVTLNEEVEV VSNEFSFKKL TCVQTRLKIF EQGLRGNFTK LKGALNMTAS YYQTYCPPTP ETDCETQVTT YADFIDSLKT FLTDIPFECK KPVQK
Indhold og opbevaring -20°C
Udtrykssystem E. coli
Form Lyophilized
Renhed eller kvalitetsklasse ≥ 98% by SDS-PAGE gel and HPLC analyses.
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.