Få mere at vide
R&D Systems™ Mouse DCIR4/CLEC4A1 (NP_955015) VersaClone cDNA
Artikelnummer. 18712118
Skift visning
Mængde:
10 μg
Artikelnummer. Mængde
18712118 10 μg
1 options
Artikelnummer. 18712118 Leverandør R&D Systems™ Leverandørnr. RDC2535
Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.
  • The Mouse DCIR4/CLEC4A1 (NP_955015) VersaClone cDNA clone contains the complete ORF for DCIR4/CLEC4A1, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The DCIR4/CLEC4A1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY
Tekniske data
Gene mDCIR4/CLEC4A1
Adgangsnummer NP_955015
Koncentration 10 μg at 0.2 μg/μL
Formulering cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gen-id (Entrez) 269799
Indhold og opbevaring Store the unopened product at -20 to -80°C
Arter Mouse
Mængde 10 μg