missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Mouse CXCL10 (IP-10) Recombinant Protein
Klik for at se tilgængelige muligheder
Mængde:
10 μg
25 μg
Beskrivelse
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
Tekniske data
Tekniske data
| Adgangsnummer | P17515 |
| Koncentration | 200 μg/mL |
| Til brug med (applikation) | Control, ELISA, Western Blot |
| Formulering | PBS with no preservative |
| Gen-id (Entrez) | 15945 |
| Molekylvægt (g/mol) | 7.8 kD |
| Navn | Mouse CXCL10 (aa 22-98) |
| Mængde | 10 μg |
| Kilde | E. coli |
| Opbevaringskrav | -20°C |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion