missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Mouse CRISP-4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5017-SP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CRISP-4 Polyclonal specifically detects CRISP-4 in Mouse samples. It is validated for Western Blot.
Tekniske data
| CRISP-4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Crisp4 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CRISP-4 Leu23-Lys250 Accession # NP_084309 | |
| 25 μg | |
| Primary | |
| Detects mouse CRISP-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse CRISP-1, -2, and -3 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| NP_084309 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10321 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion