Få mere at vide
R&D Systems™ Mouse CDO (NP_067314) VersaClone cDNA
Tekniske data
Tekniske data
| Gene | CDO |
| Adgangsnummer | NP_067314 |
| Gen-id (Entrez) | 50937 |
| Gene Alias | CDON, Cdon homolog (mouse), CDOORCAM, cell adhesion molecule-related/down-regulated by oncogenes, MGC111524, ORCAM, surface glycoprotein, Ig superfamily member |
| Indhold og opbevaring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Arter | Mouse |
| Mængde | 10 μg |