missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Mouse CD11b/Integrin alpha M Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB11241-SP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
CD11b Monoclonal specifically detects CD11b in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Tekniske data
| CD11b | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha M beta 2 Phe17-Asn1105 (Integrin alpha M), Gln24-Asn702 (Integrin beta 2) Accession # NP_032427 (Integrin alpha M) & P11835 (Integrin beta 2) | |
| 25 μg | |
| Primary | |
| Detects recombinant mouse CD11b/Integrin alpha M in Western blot. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry | |
| 908649 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells | |
| NP_032427 (Integrin alpha M) & P11835 (Integrin beta 2) | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3684 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only (RUO)
Ser du en mulighed for forbedring?Del en indholdskorrektion