missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Gibco™ Mouse BD-3, Animal-Free Recombinant Protein, PeproTech®
Beskrivelse
Recombinant Murine BD-3 is a 4.6 kDa protein containing 41 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].
Tekniske data
Tekniske data
| Til brug med (applikation) | Functional Assay |
| Format | Lyophilized |
| Formulering | protein with no preservative |
| Molekylvægt (g/mol) | 4.6 kDa |
| Navn | Mouse BD-3, Animal-Free |
| Oprensningsmetode | HPLC, SDS-PAGE |
| Mængde | 5 μg |
| Regulatorisk status | RUO |
| Endotoksin koncentration | <0.1 EU/ μg |
| Gene Alias | BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103 A; defensin, beta 103; defensin, beta 103 A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?