Få mere at vide
RAF1, Mouse, Clone: 4G4, Abnova™
Mouse monoclonal antibody raised against recombinant RAF1.
Brand: Abnova MAB10310.100ug
Beskrivelse
This gene is the cellular homolog of viral raf gene (v-raf). The encoded protein is a MAP kinase kinase kinase (MAP3K), which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated, the cellular RAF1 protein can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2, which in turn phosphorylate to activate the serine/threonine specific protein kinases, ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2. [provided by RefSeq
Tekniske data
| RAF1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. | |
| RAF1 | |
| RAF1 | |
| Recombinant protein corresponding to human RAF1. | |
| RUO | |
| 5894 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot | |
| 4G4 | |
| Mouse monoclonal antibody raised against recombinant RAF1. | |
| In PBS (0.5% protein stabilizer, 0.05% sodium azide) | |
| CRAF/NS5/Raf-1/c-Raf | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.