Få mere at vide
LTF, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native LTF.
Brand: Abnova MAB8254.100ug
Beskrivelse
This gene is a member of the transferrin family of genes and its protein product is found in the secondary granules of neutrophils. The protein is a major iron-binding protein in milk and body secretions with an antimicrobial activity, making it an important component of the nonspecific immune system. The protein demonstrates a broad spectrum of properties, including regulation of iron homeostasis, host defense against a broad range of microbial infections, anti-inflammatory activity, regulation of cellular growth and differentiation and protection against cancer development and metastasis. [provided by RefSeq
Tekniske data
| LTF | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native LTF. | |
| In PBS (0.09% sodium azide) | |
| GIG12/HLF2/LF | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts in Western blots of whole serum proteins with a lactoferrin band of 77kD. No cross-reactivity with other human serum proteins. | |
| Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| LTF | |
| LTF | |
| Native purified human LTF. | |
| RUO | |
| 4057 | |
| Human | |
| Liquid |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.