missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lidocaine, Mouse, Clone: 10-136.4, Abnova™
Mouse monoclonal antibody raised against Lidocaine.
Brand: Abnova MAB2252.100ug
Additional Details : Weight : 0.02000kg
Specifications
Lidocaine | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Radioimmune Assays (RIA) | |
10-136.4 | |
Mouse monoclonal antibody raised against Lidocaine. | |
In PBS, pH 7.4 | |
Lidocaine conjugated with KLH. | |
RUO | |
These antibodies specifically recognize lidocaine. | |
Liquid |