Få mere at vide
KIT, Mouse, Clone: 1C5, Abnova™
Mouse monoclonal antibody raised against partial recombinant KIT.
Brand: Abnova MAB10712.100uL
Beskrivelse
This gene encodes the human homolog of the proto-oncogene c-kit. C-kit was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. This protein is a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). Mutations in this gene are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous lukemia, and piebaldism. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Tekniske data
| KIT | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| KIT | |
| KIT | |
| Recombinant protein corresponding to human KIT. | |
| RUO | |
| 3815 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 1C5 | |
| Mouse monoclonal antibody raised against partial recombinant KIT. | |
| In ascites (0.03% sodium azide) | |
| C-Kit/CD117/PBT/SCFR | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.