Få mere at vide
Invitrogen™ Mouse anti-Chicken Ig Mix Secondary Antibody
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen™ SA5-10351
Beskrivelse
SA1-10645 detects chicken Immunoglobulin. Does not react with bovine, canine, donkey, equine, feline, rabbit, rat, sheep, porcine, guinea pig, goat, or human samples. SA1-10645 has been successfully used in ELISA applications. The SA1-10645 immunogen is purified chicken immunoglobulin.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Tekniske data
| Chicken Ig Mix | |
| Monoclonal | |
| 1.9 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Purified chicken immunoglobulin. | |
| 100 μg | |
| Secondary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| CIG10-183.12 | |
| Unconjugated | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Chicken | |
| Secondary Antibody | |
| IgG1 |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.