Få mere at vide
Mosapride-d5, TRC
Artikelnummer. 30473276
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30473276 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473276 Leverandør Toronto Research Chemicals Leverandørnr. TRCM7310021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mosapride-d5
CAS 1246820-66-5
Renhedsgrad noter HPLC
Molekylær formel C21H20D5ClFN3O3
Mængde 1 mg
Synonym 4-Amino-5-chloro-2-(ethoxy-d5)-N-[[4-(4-fluorophenyl)methyl]-2-morpholinyl]methyl]benzamide, Mosapid-d5, Mosid-MT-d5, Moza-d5
InChI formel InChI=1S/C21H25ClFN3O3/c1-2-28-20-10-19(24)18(22)9-17(20)21(27)25-11-16-13-26(7-8-29-16)12-14-3-5-15(23)6-4-14/h3-6,9-10,16H,2,7-8,11-13,24H2,1H3,(H,25,27)/i1D3,2D2
SMIL FC1=CC=C(CN2CC(CNC(C3=CC(Cl)=C(N)C=C3OC([2H])([2H])C([2H])([2H])[2H])=O)OCC2)C=C1
Molekylvægt (g/mol) 426.92
Procent renhed >95