Få mere at vide
Morpholine-d9, CDN
Artikelnummer. 30322270
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30322270 0.25 g
30309648 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322270 Leverandør CDN Leverandørnr. CDND59400.25G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Morpholine-D9
CAS 203578-31-8
Molekylær formel C4 D9 N O
Mængde 0.25 g
Synonym Morpholine-d9 (9 CI), 2,2,3,3,4,5,5,6,6-Nonadeuteriomorpholine, Morpholine-D9
InChI formel InChI=1 S/C4H9NO/c1-3-6-4-2-5-1/h5H,1-4H2/i1D2,2D2,3D2,4D2/hD
SMIL [2 H]N1C([2 H])([2 H])C([2 H])([2 H])OC([2 H])([2 H])C1([2 H])[2 H]
IUPAC navn 2,2,3,3,4,5,5,6,6-nonadeuteriomorpholine
Molekylvægt (g/mol) 96.176
Formel vægt 96.125 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity