Få mere at vide
Morpholine-d8, TRC
Artikelnummer. 30475775
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30475775 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475775 Leverandør Toronto Research Chemicals Leverandørnr. TRCM72372825MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Morpholine D8
CAS 342611-02-3
Molekylær formel C4 2H8 H N O
Mængde 25 mg
Synonym Morpholine-2,2,3,3,5,5,6,6-d8
InChI formel InChI=1S/C4H9NO/c1-3-6-4-2-5-1/h5H,1-4H2/i1D2,2D2,3D2,4D2
SMIL [2H]C1([2H])NC([2H])([2H])C([2H])([2H])OC1([2H])[2H]
IUPAC navn 2,2,3,3,5,5,6,6-octadeuteriomorpholine
Molekylvægt (g/mol) 95.17
Formel vægt 95.1186