missing translation for 'onlineSavingsMsg'
Learn More

morpholine-4-carboximidamide hydroiodide, Thermo Scientific™

Catalog Number p-4391437
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
102392-87-0
C5H12IN3O
257.075
LJQSGLRGQQZIMF-UHFFFAOYSA-N
morpholine-4-carboximidamide hydroiodide, c5h11n3o.hi, morpholine-4-carboxamidine, iodide, morpholine-4-carboxamidine hydroiodide
2778396
morpholine-4-carboximidamide;hydroiodide
C1COCCN1C(=N)N.I
This item is not returnable. View return policy
102392-87-0
C5H12IN3O
257.075
LJQSGLRGQQZIMF-UHFFFAOYSA-N
morpholine-4-carboximidamide hydroiodide, c5h11n3o.hi, morpholine-4-carboxamidine, iodide, morpholine-4-carboxamidine hydroiodide
2778396
morpholine-4-carboximidamide;hydroiodide
C1COCCN1C(=N)N.I
Quantity 250 mg
Formula Weight 257.07
Percent Purity Tech.
Grade Technical
Chemical Name or Material Morpholine-4-carboximidamide hydroiodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'