missing translation for 'onlineSavingsMsg'
Få mere at vide

Morinidazole, MedChemExpress

Artikelnummer. 16459377
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
Pakningsstørrelse:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16459377 5 mg 5mg
16416838 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16459377 Leverandør Medchem Express Leverandørnr. HY15781

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Vea la política de devoluciones

MedChemExpress Morinidazole is a novel 5-nitroimidazole antimicrobial drug that undergoes extensive metabolism in humans via N+-glucuronidation and sulfation, for the treatment of bacterial infections including appendicitis and pelvic inflammatory disease (PID) caused by anaerobic bacteria.

Morinidazole is a novel 5-nitroimidazole antimicrobial drug that undergoes extensive metabolism in humans via N+-glucuronidation and sulfation, for the treatment of bacterial infections including appendicitis and pelvic inflammatory disease (PID) caused by anaerobic bacteria.

Especificaciones

Kemisk navn eller materiale Morinidazole
Fysisk form Solid
CAS 92478-27-8
Farve Off-White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C11H18N4O4
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation DMSO : ≥ 100 mg/mL (369.97 mM)
SMIL OC(CN1C([N+]([O-])=O)=CN=C1C)CN2CCOCC2
Molekylvægt (g/mol) 270.29
Formel vægt 270.29
Procent renhed 98.05%
Grad Research
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.