Få mere at vide
Morinamide Hydrochloride, Mikromol™
Beskrivelse
Morinamide Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Morinamide Hydrochloride |
| CAS | 1473-73-0 |
| Analyt- eller komponentnavne | Morinamide Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C10 H14 N4 O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 2-Pyrazinecarboxamide, N-(4-morpholinylmethyl)-, hydrochloride (1:1), Pyrazinecarboxamide, N-(4-morpholinylmethyl)-, monohydrochloride (9CI), Pyrazinecarboxamide, N-(morpholinomethyl)-, monohydrochloride (8CI), B 2311, Morinamide hydrochloride, Morphazinamide hydrochloride, Piazofolina |
| InChI formel | InChI=1S/C10H14N4O2.ClH/c15-10(9-7-11-1-2-12-9)13-8-14-3-5-16-6-4-14;/h1-2,7H,3-6,8H2,(H,13,15);1H |
| SMIL | Cl.O=C(NCN1CCOCC1)c2cnccn2 |
| IUPAC navn | N-(morpholin-4-ylmethyl)pyrazine-2-carboxamide;hydrochloride |
| Vis mere |