Få mere at vide
Morinamide Hydrochloride, LoGiCal
Artikelnummer. 30458558
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30458558 10 mg
1 options
Artikelnummer. 30458558 Leverandør LoGiCal Leverandørnr. LGCFOR3142.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Morinamide Hydrochloride
CAS 1473-73-0
Analyt- eller komponentnavne Morinamide Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C10 H14 N4 O2 . Cl H
Certificeringer/compliance ISO 9001
Synonym 2-Pyrazinecarboxamide, N-(4-morpholinylmethyl)-, hydrochloride (1:1), Pyrazinecarboxamide, N-(4-morpholinylmethyl)-, monohydrochloride (9CI), Pyrazinecarboxamide, N-(morpholinomethyl)-, monohydrochloride (8CI), B 2311, Morinamide hydrochloride, Morphazinamide hydrochloride, Piazofolina
InChI formel InChI=1S/C10H14N4O2.ClH/c15-10(9-7-11-1-2-12-9)13-8-14-3-5-16-6-4-14;/h1-2,7H,3-6,8H2,(H,13,15);1H
Løsningstype Neat
SMIL Cl.O=C(NCN1CCOCC1)c2cnccn2
IUPAC navn N-(morpholin-4-ylmethyl)pyrazine-2-carboxamide;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 258.7
Mængde 10 mg
Formel vægt 258.0884 g/mol