Få mere at vide
Montelukast Sulfoxide (Mixture of Diastereomers), TRC
Artikelnummer. 17999154
Skift visning
Mængde:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17999154 1 mg
17957344 50 mg
17928694 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17999154 Leverandør Toronto Research Chemicals Leverandørnr. TRCM5680201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1152185-58-4
Molekylær formel C35 H36 Cl N O4 S
Molekylvægt (g/mol) 602.18
Synonym 1-[[[1-[3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]sulfinyl]methyl]cyclopropaneacetic Acid, Montelukast Sulfoxide, Cyclopropaneacetic acid, 1-[[[(1R)-1-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]sulfinyl]methyl]-, 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]sulfinyl]methyl]cyclopropaneacetic acid, Montelukast sulfoxide, [1-[[[(1R)-1-[3-[(E)-2-(7-Chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]sulfinyl]methyl]cyclopropyl]acetic acid, 1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]sulfinyl]methyl]cyclopropaneacetic Acid, Montelukast M2, Montelukast EP Impurity C, Montelukast USP Related Compound C
IUPAC navn 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloro-2-quinolyl)vinyl]phenyl]-3-[2-(1-hydroxy-1-methyl-ethyl)phenyl]propyl]sulfinylmethyl]cyclopropyl]acetic acid
SMIL CC(C)(O)c1ccccc1CC[C@H](c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2)S(=O)CC5(CC(=O)O)CC5
Tekniske data
Kemisk navn eller materiale Montelukast Sulfoxide
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C35H36ClNO4S/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(42(41)23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29/h3-12,14-16,20-21,32,40H,13,17-19,22-23H2,1-2H3,(H,38,39)/b15-10+/t32-,42?/m1/s1
Formel vægt 601.2054
Procent renhed >95%