missing translation for 'onlineSavingsMsg'
Learn More

Monosulfuron, TRC

Product Code. 30473261
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30473261 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30473261 Supplier Toronto Research Chemicals Supplier No. TRCM567185100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Monosulfuron
CAS 155860-63-2
Molekylær formel C12 H11 N5 O5 S
Mængde 100 mg
Synonym Benzenesulfonamide, N-[[(4-methyl-2-pyrimidinyl)amino]carbonyl]-2-nitro-, Monosulfuron
InChI formel InChI=1S/C12H11N5O5S/c1-8-6-7-13-11(14-8)15-12(18)16-23(21,22)10-5-3-2-4-9(10)17(19)20/h2-7H,1H3,(H2,13,14,15,16,18)
SMIL Cc1ccnc(NC(=O)NS(=O)(=O)c2ccccc2[N+](=O)[O-])n1
IUPAC navn 1-(4-methylpyrimidin-2-yl)-3-(2-nitrophenyl)sulfonylurea
Molekylvægt (g/mol) 337.31
Formel vægt 337.0481
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.