Få mere at vide
Monosodium Acid Methane Arsonate Sesquihydrate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methylarsonic acid sodium |
| CAS | 2163-80-6 |
| Molekylær formel | C H4 As O3 . Na |
| Mængde | 100 mg |
| Synonym | Methanearsonic acid, monosodium salt (8CI), Ansar 170, Ansar 170L, Ansar 529, Bueno 6, Daconate, Gepiron, MSMA, Monosodium acid methanearsonate, Monosodium acid metharsonate, Monosodium methanearsonate, Monosodium methylarsonate, Sodium acid methanearsonate, Weed 108 |
| InChI formel | InChI=1S/CH5AsO3.Na/c1-2(3,4)5;/h1H3,(H2,3,4,5);/q;+1/p-1 |
| SMIL | [Na+].C[As](=O)(O)[O-] |
| IUPAC navn | sodium;hydroxy(methyl)arsinate |
| Molekylvægt (g/mol) | 161.952 |
| Formel vægt | 161.9274 |