Få mere at vide
Monopropylheptylphthalate 6-Hydroxy-d4, TRC
Artikelnummer. 30471966
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30471966 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30471966 Leverandør Toronto Research Chemicals Leverandørnr. TRCM5670771MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Monopropylheptylphthalate 6-Hydroxy-d4
CAS 1412411-11-0
Molekylær formel C18H22D4O5
Mængde 1 mg
Synonym OH-MiDP-d4, 1,2-Benzene-3,4,5,6-d4-dicarboxylic Acid 1-(6-Hydroxy-2-propylheptyl) Ester, OH-MPHP-d4
InChI formel InChI=1S/C18H26O5/c1-3-7-14(9-6-8-13(2)19)12-23-18(22)16-11-5-4-10-15(16)17(20)21/h4-5,10-11,13-14,19H,3,6-9,12H2,1-2H3,(H,20,21)
SMIL OC(C1=C([2H])C([2H])=C([2H])C([2H])=C1C(OCC(CCC)CCCC(O)C)=O)=O
Molekylvægt (g/mol) 326.42