Få mere at vide
Monopropyl Phthalate-d4, TRC
Artikelnummer. 30475425
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475425 5 mg
30479959 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30475425 Leverandør Toronto Research Chemicals Leverandørnr. TRCM5671025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1795020-91-5
Molekylær formel C11 D4 H8 O4
Molekylvægt (g/mol) 212.235
Synonym 2,3,4,5-Tetradeuterio-6-propoxycarbonylbenzoic acid, mono-Propyl Phthalate-3,4,5,6-d4, Monopropyl Phthalate-d4, 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid monopropyl ester, Phthalic acid-d4 monopropyl ester, Propyl hydrogen phthalate-d4, Monopropyl Phthalate-3,4,5,6-D4
IUPAC navn 2,3,4,5-tetradeuterio-6-propoxycarbonylbenzoic acid
SMIL [2H]c1c([2H])c([2H])c(C(=O)OCCC)c(C(=O)O)c1[2H]
Tekniske data
Kemisk navn eller materiale Monopropyl Phthalate-d4
Mængde 5 mg
InChI formel InChI=1S/C11H12O4/c1-2-7-15-11(14)9-6-4-3-5-8(9)10(12)13/h3-6H,2,7H2,1H3,(H,12,13)/i3D,4D,5D,6D
Formel vægt 212.099