missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Monomethylsuberat, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Monomethyl Suberate |
| CAS | 3946-32-5 |
| Molekylær formel | C9H16O4 |
| Mængde | 5 g |
| Synonym | Octanedioic Acid, Monomethyl Ester, Suberic Acid Methyl Ester, Suberic Acid, Monomethyl Ester, 8-Methoxy-8-oxooctanoic Acid, Methyl 1,8-Octanedionate, Methyl 7-Carboxyheptanoate |
| InChI formel | InChI=1S/C9H16O4/c1-13-9(12)7-5-3-2-4-6-8(10)11/h2-7H2,1H3,(H,10,11) |
| SMIL | COC(=O)CCCCCCC(=O)O |
| IUPAC navn | 8-methoxy-8-oxooctanolsyre |
| Molekylvægt (g/mol) | 188.22 |
| Formel vægt | 188.1049 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?