Få mere at vide
Monomethyl Glutarate, TRC
Artikelnummer. 30479010
Skift visning
Mængde:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479010 25 g
30468835 100 g
30474687 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30479010 Leverandør Toronto Research Chemicals Leverandørnr. TRCM56650025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1501-27-5
Molekylær formel C6H10O4
Molekylvægt (g/mol) 146.14
Synonym 4-(Methoxycarbonyl)butyric acid, 4-Carboxybutanoic acid methyl ester4-Methoxycarbonylbutanoic acid, 5-Methoxy-5-oxopentanoic acid, Glutaric acid methyl half ester, Monomethyl pentanedioate, NSC 93807
IUPAC navn 5-methoxy-5-oxopentanoic acid
SMIL COC(=O)CCCC(=O)O
Tekniske data
Kemisk navn eller materiale Monomethyl Glutarate
Mængde 25 g
InChI formel InChI=1S/C6H10O4/c1-10-6(9)4-2-3-5(7)8/h2-4H2,1H3,(H,7,8)
Formel vægt 146.0579