Få mere at vide
Monoisopropyl Phthalate-d4, TRC
Artikelnummer. 30481506
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481506 10 mg
30472038 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30481506 Leverandør Toronto Research Chemicals Leverandørnr. TRCM54790210MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2733160-00-2
Molekylær formel C11 D4 H8 O4
Molekylvægt (g/mol) 212.2352
Synonym mono-iso-Propyl Phthalate-3,4,5,6-d4, Monoisopropyl Phthalate-d4, Phthalic Acid Monoisopropyl Ester-d4, Phthalic Acid-d4 Monoisopropyl Ester, Monoisopropyl Phthalate-3,4,5,6-D4
IUPAC navn 2,3,4,5-tetradeuterio-6-propan-2-yloxycarbonylbenzoic acid
SMIL [2H]c1c([2H])c([2H])c(C(=O)OC(C)C)c(C(=O)O)c1[2H]
Tekniske data
Kemisk navn eller materiale Monoisopropyl Phthalate-d4
Mængde 10 mg
InChI formel InChI=1S/C11H12O4/c1-7(2)15-11(14)9-6-4-3-5-8(9)10(12)13/h3-7H,1-2H3,(H,12,13)/i3D,4D,5D,6D
Formel vægt 212.0987