missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Monoethyl Fumarate Magnesium, Mikromol™
Beskrivelse
Monoethyl Fumarate Magnesium
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Monoethyl Fumarate Magnesium |
| CAS | 83918-60-9 |
| Analyt- eller komponentnavne | Monoethyl Fumarate Magnesium |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | 2 C6 H7 O4 . Mg |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Monoethyl Fumarate Magnesium, 2-Butenedioic acid (2E)-, monoethyl ester, magnesium salt (9CI), 2-Butenedioic acid (E)-, monoethyl ester, magnesium salt, 2-Butenedioic acid (2E)-, 1-ethyl ester, magnesium salt (2:1) |
| InChI formel | InChI=1S/2C6H8O4.Mg/c2*1-2-10-6(9)4-3-5(7)8;/h2*3-4H,2H2,1H3,(H,7,8);/q;;+2/p-2/b2*4-3+; |
| SMIL | [Mg+2].CCOC(=O)\C=C\C(=O)[O-].CCOC(=O)\C=C\C(=O)[O-] |
| IUPAC navn | magnesium;(E)-4-ethoxy-4-oxobut-2-enoate |
| Vis mere |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?