Få mere at vide
Monoethyl Fumarate Magnesium, LoGiCal
Artikelnummer. 30459422
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459422 10 mg
1 options
Artikelnummer. 30459422 Leverandør LoGiCal Leverandørnr. LGCFOR3481.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Monoethyl Fumarate Magnesium
CAS 83918-60-9
Analyt- eller komponentnavne Monoethyl Fumarate Magnesium
Forsendelsestilstand Room Temperature
Molekylær formel 2 C6 H7 O4 . Mg
Certificeringer/compliance ISO 9001
Synonym Monoethyl Fumarate Magnesium, 2-Butenedioic acid (2E)-, monoethyl ester, magnesium salt (9CI), 2-Butenedioic acid (E)-, monoethyl ester, magnesium salt, 2-Butenedioic acid (2E)-, 1-ethyl ester, magnesium salt (2:1)
InChI formel InChI=1S/2C6H8O4.Mg/c2*1-2-10-6(9)4-3-5(7)8;/h2*3-4H,2H2,1H3,(H,7,8);/q;;+2/p-2/b2*4-3+;
Løsningstype Neat
SMIL [Mg+2].CCOC(=O)\C=C\C(=O)[O-].CCOC(=O)\C=C\C(=O)[O-]
IUPAC navn magnesium;(E)-4-ethoxy-4-oxobut-2-enoate
Industry Type Clinical Testing
Molekylvægt (g/mol) 310.54
Mængde 10 mg
Formel vægt 310.0539 g/mol