Få mere at vide
Monoethanolamine Standard for Ion Chromatography, 1000 mg/L in water, Honeywell Fluka™
Shop alle Honeywell Fluka produkterBeskrivelse
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Monoethanolamine Standard for Ion Chromatography |
| Tæthed | 1.01 g/cm3 |
| Mængde | 100 ml |
| Grad | Standards for Ion Chromatography |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.