missing translation for 'onlineSavingsMsg'
Få mere at vide

Monocyclohexyl Phthalate-d4, TRC

Artikelnummer. 30482058
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482058 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482058 Leverandør Toronto Research Chemicals Leverandørnr. TRCM5266025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Monocyclohexyl Phthalate-d4
CAS 1398066-18-6
Molekylær formel C14 D4 H12 O4
Mængde 5 mg
Synonym mono-Cyclohexyl Phthalate-3,4,5,6-d4, Monocyclohexyl Phthalate-d4, 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid monocyclohexyl ester, Phthalic acid-d4 monocyclohexyl ester, Cyclohexyl hydrogen phthalate-d4, Monocyclohexyl Phthalate-3,4,5,6-D4
InChI formel InChI=1S/C14H16O4/c15-13(16)11-8-4-5-9-12(11)14(17)18-10-6-2-1-3-7-10/h4-5,8-10H,1-3,6-7H2,(H,15,16)/i4D,5D,8D,9D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OC2CCCCC2)c(C(=O)O)c1[2H]
IUPAC navn 2-cyclohexyloxycarbonyl-3,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 252.299
Formel vægt 252.13
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.