Få mere at vide
Monocrotophos, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Monocrotophos |
| CAS | 6923-22-4 |
| Molekylær formel | C7 H14 N O5 P |
| Mængde | 100 mg |
| Synonym | Phosphoric acid, dimethyl (1E)-1-methyl-3-(methylamino)-3-oxo-1-propenyl ester (9CI), Phosphoric acid, dimethyl 1-methyl-3-(methylamino)-3-oxo-1-propenyl ester, (E)-, Phosphoric acid, dimethyl ester, ester with 3-hydroxy-N-methylcrotonamide, (E)- (8CI), 3-Hydroxy-N-methyl-cis-crotonamide dimethyl phosphate, Apadrin, Azodrin, Azodrin 202R, Azodrin 60, C 1414, Corophos, Croton 36, Devi, Kadett, Kadett 36, Monicil, Monochrotophos, Monocil, Monocil 40, Monocos, Monocron, Monocrotophos, Monodhan, Monokrotofos, Monostar, Nuvacron, Nuvacron 20, Nuvacron 36SL, Parryfos, Phytofos, SD 9129, Sufos |
| InChI formel | InChI=1S/C7H14NO5P/c1-6(5-7(9)8-2)13-14(10,11-3)12-4/h5H,1-4H3,(H,8,9)/b6-5+ |
| SMIL | CNC(=O)\C=C(/C)\OP(=O)(OC)OC |
| IUPAC navn | dimethyl [(E)-4-(methylamino)-4-oxobut-2-en-2-yl] phosphate |
| Molekylvægt (g/mol) | 223.16 |
| Formel vægt | 223.061 |