Få mere at vide
Monocaprylin, TRC
Artikelnummer. 30483749
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483749 1 g
30468333 250 mg
30471829 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483749 Leverandør Toronto Research Chemicals Leverandørnr. TRCM5672531G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 502-54-5
Molekylær formel C11H22O4
Molekylvægt (g/mol) 218.29
Synonym Capmul MCM-C 8, Capmul MCM-L 8, Caprylic acid monoglyceride, Drewmulse CA, Glycerin Monocaprylate, Glycerin Monooctanoate, Glycerol Monocaprylate, Glycerol Monooctanoate, Glyceryl Monocaprylate, Glyceryl Monooctanoate, Homotex PT
IUPAC navn 2,3-dihydroxypropyl octanoate
SMIL CCCCCCCC(=O)OCC(CO)O
Tekniske data
Kemisk navn eller materiale Monocaprylin
Mængde 1 g
InChI formel InChI=1S/C11H22O4/c1-2-3-4-5-6-7-11(14)15-9-10(13)8-12/h10,12-13H,2-9H2,1H3
Formel vægt 218.15