Få mere at vide
Monobutyl Phosphate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Monobutyl Phosphate |
| CAS | 1623-15-0 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C4H11O4P |
| Mængde | 100 mg |
| Synonym | Phosphoric Acid Monobutyl Ester, Butyl Diacid Phosphate, Butyl Dibasic Acid Phosphate, Butyl Dihydrogen Phosphate, Dihydrogen Monobutyl Phosphate, JAMP 4P, MP 4, Monobutyl Dihydrogen Phosphate, Monobutyl Phosphate, NSC 41911 |
| InChI formel | InChI=1S/C4H11O4P/c1-2-3-4-8-9(5,6)7/h2-4H2,1H3,(H2,5,6,7) |
| SMIL | CCCCOP(=O)(O)O |
| IUPAC navn | butyl dihydrogen phosphate |
| Molekylvægt (g/mol) | 154.1 |
| Vis mere |