Få mere at vide
Monobutyl Phosphate-d9, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Monobutyl Phosphate-d9 |
| CAS | 156213-20-6 |
| Molekylær formel | C4H2D9O4P |
| Mængde | 25 mg |
| Synonym | Phosphoric Acid Monobutyl-d9 Ester, Butyl-d9 Diacid Phosphate, Butyl-d9 Dibasic Acid Phosphate, Butyl-d9 Dihydrogen Phosphate, Dihydrogen Monobutyl-d9 Phosphate, JAMP 4P-d9, MP 4-d9, Monobutyl-d9 Dihydrogen Phosphate, Monobutyl-d9 Phosphate, NSC 41911-d9 |
| InChI formel | InChI=1S/C4H11O4P/c1-2-3-4-8-9(5,6)7/h2-4H2,1H3,(H2,5,6,7)/i1D3,2D2,3D2,4D2 |
| SMIL | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OP(=O)(O)O |
| Molekylvægt (g/mol) | 163.16 |