missing translation for 'onlineSavingsMsg'
Få mere at vide

Monobutyl Phosphate Biscyclohexylamine Salt, TRC

Artikelnummer. 30468564
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30468564 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30468564 Leverandør Toronto Research Chemicals Leverandørnr. TRCM52505510G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Monobutyl Phosphate Biscyclohexylamine Salt
CAS 35314-60-4
Molekylær formel C6 H14 N . C4 H9 O4 P
Mængde 10 g
Synonym Phosphoric Acid Monobutyl Ester Biscyclohexylamine Salt, Butyl Diacid Phosphate Biscyclohexylamine Salt, Butyl Dibasic Acid Phosphate Biscyclohexylamine Salt, Butyl Dihydrogen Phosphate Biscyclohexylamine Salt, Dihydrogen Monobutyl Phosphate Biscyclohexylamine Salt, JAMP 4P Biscyclohexylamine Salt, MP 4 Biscyclohexylamine Salt, Monobutyl Dihydrogen Phosphate Biscyclohexylamine Salt, Monobutyl Phosphate Biscyclohexylamine Salt, NSC 41911 Biscyclohexylamine Salt
InChI formel InChI=1S/C6H13N.C4H11O4P/c7-6-4-2-1-3-5-6;1-2-3-4-8-9(5,6)7/h6H,1-5,7H2;2-4H2,1H3,(H2,5,6,7)/p-1
SMIL CCCCOP(=O)([O-])[O-].[NH3+]C1CCCCC1
IUPAC navn butyl phosphate;cyclohexylazanium
Molekylvægt (g/mol) 252.268
Formel vægt 252.137
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.