Få mere at vide
mono-n-Undecyl Phthalate-3,4,5,6-d4, CDN
Artikelnummer. 30320045
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320045 0.1 g 0.1g
30313798 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320045 Leverandør CDN Leverandørnr. CDND77040.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Monoundecyl Phthalate-3,4,5,6-D4
CAS 2733718-42-6
Molekylær formel C19 D4 H24 O4
Mængde 0.1 g
Synonym mono-n-Undecyl phthalate-3,4,5,6-d4, 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid 1-undecyl ester, Hydrogen undecyl phthalate-d4, Monoundecyl phthalate-3,4,5,6-d4, Phthalic acid-d4 monoundecyl ester, Monoundecyl phthalate-3,4,5,6-D4
InChI formel InChI=1 S/C19H28O4/c1-2-3-4-5-6-7-8-9-12-15-23-19(22)17-14-11-10-13-16(17)18(20)21/h10-11,13-14 H,2-9,12,15H2,1H3,(H,20,21)/i10D,11 D,13 D,14 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCCCCCC)c(C(=O)O)c1[2 H]
IUPAC navn 2,3,4,5-tetradeuterio-6-undecoxycarbonylbenzoic acid
Molekylvægt (g/mol) 324.448
Formel vægt 324.224 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity