missing translation for 'onlineSavingsMsg'
Få mere at vide

mono-Cyclohexyl Phthalate-3,4,5,6-d4, CDN

Artikelnummer. 30326612
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30326612 0.1 g 0.1g
30317390 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326612 Leverandør CDN Leverandørnr. CDND74890.1G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Monocyclohexyl Phthalate-d4
CAS 1398066-18-6
Molekylær formel C14 D4 H12 O4
Mængde 0.1 g
Synonym mono-Cyclohexyl Phthalate-3,4,5,6-d4, Monocyclohexyl Phthalate-d4, 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid monocyclohexyl ester, Phthalic acid-d4 monocyclohexyl ester, Cyclohexyl hydrogen phthalate-d4, Monocyclohexyl Phthalate-3,4,5,6-D4
InChI formel InChI=1 S/C14H16O4/c15-13(16)11-8-4-5-9-12(11)14(17)18-10-6-2-1-3-7-10/h4-5,8-10 H,1-3,6-7H2,(H,15,16)/i4D,5 D,8 D,9 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OC2CCCCC2)c(C(=O)O)c1[2 H]
IUPAC navn 2-cyclohexyloxycarbonyl-3,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 252.299
Formel vægt 252.13 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.