Få mere at vide
Mono(3-methylpentyl) Phthalate, TRC
Artikelnummer. 30470793
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470793 250 mg
30466887 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30470793 Leverandør Toronto Research Chemicals Leverandørnr. TRCM566615250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 106636-65-1
Molekylær formel C14H18O4
Molekylvægt (g/mol) 250.29
Synonym 1,2-Benzenedicarboxylic Acid 1-(3-Methylpentyl) Ester, 1,2-Benzenedicarboxylic Acid Mono(3-methylpentyl) Ester
IUPAC navn 2-(3-methylpentoxycarbonyl)benzoic acid
SMIL CCC(C)CCOC(=O)c1ccccc1C(=O)O
Tekniske data
Kemisk navn eller materiale Mono(3-methylpentyl) Phthalate
Mængde 250 mg
InChI formel InChI=1S/C14H18O4/c1-3-10(2)8-9-18-14(17)12-7-5-4-6-11(12)13(15)16/h4-7,10H,3,8-9H2,1-2H3,(H,15,16)
Formel vægt 250.1205