Få mere at vide
Mono[2-(carboxymethyl)hexyl] Phthalate-d4, TRC
Artikelnummer. 30468109
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30468109 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30468109 Leverandør Toronto Research Chemicals Leverandørnr. TRCM5255871MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Mono[2-(carboxymethyl)hexyl] Phthalate-d4
CAS 1794811-42-9
Renhedsgrad noter HPLC
Molekylær formel C16H16D4O6
Mængde 1 mg
Synonym 1,2-(Benzene-d4)dicarboxylic Acid 1-[2-(Carboxymethyl)hexyl] Ester, MCMHP-d4, 1,2-(Benzene-d4)dicarboxylic Acid Mono[2-(carboxymethyl)hexyl] Ester
InChI formel InChI=1S/C16H20O6/c1-2-3-6-11(9-14(17)18)10-22-16(21)13-8-5-4-7-12(13)15(19)20/h4-5,7-8,11H,2-3,6,9-10H2,1H3,(H,17,18)(H,19,20)/i4D,5D,7D,8D
SMIL O=C(OCC(CC(O)=O)CCCC)C1=C([2H])C([2H])=C([2H])C([2H])=C1C(O)=O
IUPAC navn 2-(((2-(carboxymethyl)hexyl)oxy)carbonyl)benzoic-3,4,5,6-d4 acid
Molekylvægt (g/mol) 312.35
Formel vægt 312.15
Procent renhed >95