Learn More
Molybdic Acid About 85% MoO3, Supelco™
Product Code. 15833880
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Product Code. Quantity Unit Size
15833880 1 kg 1kg
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15833880 Supplier Supelco Supplier No. 1004001000
 Restricted Item
Forventet forsendelse: 19-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
Containing ammonium molybdate EMPLURA™
(containing ammonium molybdate) EMPLURA™
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 300°C
CAS 7782-91-4
Quantity 1 kg
MDL Number MFCD00036287
Synonym Molybdic acid about 85% MoO3, Molybdic acid about 85% MoO3
Molecular Weight (g/mol) 161.95
Grade Research
Density 3.1 g/cm3
Chemical Name or Material Molybdic Acid About 85% MoO3